siRNA Details
| Gene Symbol: CGGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008390.1 | NP_001008391.1 | 4 | 874 | ||
| NM_001195308.1 | NP_001182237.1 | 4 | 855 | |||
| NM_003663.3 | NP_003654.3 | 3 | 772 | |||
| XM_017007355.1 | XP_016862844.1 | 3 | 680 | |||
| GenBank mRNA | AJ000258.1 | - | 3 | 558 | ||
| AL117392.1 | - | 3 | 718 | |||
| BC052980.1 | - | 3 | 674 | 6012191 | ||
| CR456854.1 | - | 1 | 362 | |||

CGGBP1
Genomic Map