siRNA Details
| Gene Symbol: Paqr7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034081.1 | NP_001029253.1 | 1 | 190 | ||
| XM_006239130.2 | XP_006239192.1 | 2 | 288 | |||
| XM_008764145.1 | XP_008762367.1 | 3 | 511 | |||
| XM_008764146.1 | XP_008762368.1 | 3 | 299 | |||
| GenBank mRNA | DQ027002.1 | - | 1 | 190 | ||
More Information
Set Membership: |
|

Paqr7
Genomic Map