siRNA Details
| Gene Symbol: Tmem57 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025699.2 | NP_001020870.2 | 6 | 1048 | ||
| XM_008764240.1 | XP_008762462.1 | 5 | 1244 | |||
| XM_008764241.1 | XP_008762463.1 | 6 | 1000 | |||
| GenBank mRNA | AY845021.1 | - | 6 | 1048 | ||
| BC097999.1 | - | 6 | 974 | 7453136 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_025382, AY846873, BC054769, AY364165, AK138874, and others.

Tmem57
Genomic Map