siRNA Details
| Gene Symbol: Hp1bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199108.1 | NP_954539.1 | 12 | 1837 | ||
| GenBank mRNA | BC061837.1 | - | 12 | 1837 | 5599940 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hp1bp3| Gene Symbol: Hp1bp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199108.1 | NP_954539.1 | 12 | 1837 | ||
| GenBank mRNA | BC061837.1 | - | 12 | 1837 | 5599940 | |