siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 7, 8 | 802 | ||
| GenBank mRNA | BC082075.1 | - | 7, 8 | 802 | 7133207 | |
| FQ214046.1 | - | 2, 3 | 248 | |||
| FQ228905.1 | - | 7, 8 | 834 | |||
| FQ229122.1 | - | 7, 8 | 814 | |||

Ddost
Genomic Map