siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 2 | 212 | ||
| GenBank mRNA | BC082075.1 | - | 2 | 212 | 7133207 | |
| FQ228905.1 | - | 2 | 244 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddost| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 2 | 212 | ||
| GenBank mRNA | BC082075.1 | - | 2 | 212 | 7133207 | |
| FQ228905.1 | - | 2 | 244 | |||