siRNA Details
| Gene Symbol: Ddost | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012104.1 | NP_001012104.1 | 9 | 1025 | ||
| GenBank mRNA | BC082075.1 | - | 9 | 1025 | 7133207 | |
| FQ214046.1 | - | 4 | 471 | |||
| FQ228905.1 | - | 9 | 1057 | |||
| FQ229122.1 | - | 9 | 1037 | |||

Ddost
Genomic Map