siRNA Details
| Gene Symbol: Vwa5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107988.1 | NP_001101458.1 | 17 | 2803 | ||
| XM_006239199.2 | XP_006239261.1 | 17 | 3517 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vwa5b1| Gene Symbol: Vwa5b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107988.1 | NP_001101458.1 | 17 | 2803 | ||
| XM_006239199.2 | XP_006239261.1 | 17 | 3517 | |||