siRNA Details
| Gene Symbol: Dhrs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037199.3 | NP_001032276.3 | 4 | 951 | ||
| GenBank mRNA | BC107450.1 | - | 4 | 951 | 7322355 | |
| EF125189.1 | - | 4 | 1397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dhrs3| Gene Symbol: Dhrs3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037199.3 | NP_001032276.3 | 4 | 951 | ||
| GenBank mRNA | BC107450.1 | - | 4 | 951 | 7322355 | |
| EF125189.1 | - | 4 | 1397 | |||