siRNA Details
| Gene Symbol: Spsb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107994.2 | NP_001101464.1 | 2 | 724 | ||
| XM_006239462.2 | XP_006239524.1 | 2 | 757 | |||
| XM_008764257.1 | XP_008762479.1 | 2 | 970 | |||
| XM_008764258.1 | XP_008762480.1 | 1 | 555 | |||
| XM_008764259.1 | XP_008762481.1 | 2 | 2701 | |||
| GenBank mRNA | BC158603.1 | - | 2 | 724 | 7133644 | |

Spsb1
Genomic Map