siRNA Details
| Gene Symbol: Gpr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012107.1 | NP_001012107.1 | 2 | 661 | ||
| GenBank mRNA | BC090013.1 | - | 2 | 661 | 7324631 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gpr157| Gene Symbol: Gpr157 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012107.1 | NP_001012107.1 | 2 | 661 | ||
| GenBank mRNA | BC090013.1 | - | 2 | 661 | 7324631 | |
Set Membership: |
|