siRNA Details
| Gene Symbol: Errfi1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001014071.1 | NP_001014093.1 | 4 | 960 | ||
| XM_008764260.1 | XP_008762482.1 | 3 | 3708 | |||
| XM_008764261.1 | XP_008762483.1 | 4 | 1044 | |||
| XM_008764262.1 | XP_008762484.1 | 3 | 3479 | |||
| GenBank mRNA | BC083845.1 | - | 4 | 960 | 7121187 | |
| X07266.1 | - | 4 | 963 | |||

Errfi1
Genomic Map