siRNA Details
| Gene Symbol: Klhl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107996.1 | NP_001101466.1 | 2, 3 | 1420 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl21| Gene Symbol: Klhl21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107996.1 | NP_001101466.1 | 2, 3 | 1420 | ||