siRNA Details
| Gene Symbol: Nphp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037650.2 | NP_001032739.2 | 15 | 2219 | ||
| XM_006239464.2 | XP_006239526.1 | 15 | 9891 | |||
| XM_006239465.2 | XP_006239527.1 | 14 | 1931 | |||
| XM_006239467.2 | XP_006239529.1 | 15 | 3276 | |||
| XM_008764358.1 | XP_008762580.1 | 16 | 3406 | |||
| XM_008764359.1 | XP_008762581.1 | 15 | 9891 | |||

Nphp4
Genomic Map