siRNA Details
| Gene Symbol: Plch2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008764393.1 | XP_008762615.1 | 6 | 1917 | ||
| XM_008764394.1 | XP_008762616.1 | 6 | 974 | |||
| XM_008764395.1 | XP_008762617.1 | 6 | 812 | |||
| XM_008764396.1 | XP_008762618.1 | 6 | 965 | |||
| XM_008764397.1 | XP_008762619.1 | 7 | 988 | |||
| XM_008764398.1 | XP_008762620.1 | 6 | 1916 | |||

Plch2
Genomic Map