siRNA Details
| Gene Symbol: Cptp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007703.1 | NP_001007704.1 | 3 | 747 | ||
| XM_006239576.2 | XP_006239638.1 | 3 | 908 | |||
| GenBank mRNA | BC083599.1 | - | 3 | 747 | 7133008 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cptp| Gene Symbol: Cptp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007703.1 | NP_001007704.1 | 3 | 747 | ||
| XM_006239576.2 | XP_006239638.1 | 3 | 908 | |||
| GenBank mRNA | BC083599.1 | - | 3 | 747 | 7133008 | |