siRNA Details
| Gene Symbol: Acap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107999.1 | NP_001101469.1 | 15 | 1777 | ||
| XM_006239577.2 | XP_006239639.1 | 15 | 1313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Acap3| Gene Symbol: Acap3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107999.1 | NP_001101469.1 | 15 | 1777 | ||
| XM_006239577.2 | XP_006239639.1 | 15 | 1313 | |||