siRNA Details
| Gene Symbol: Rnf19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108003.1 | NP_001101473.1 | 3 | 441 | ||
| XM_008764147.1 | XP_008762369.1 | 5 | 1008 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rnf19b| Gene Symbol: Rnf19b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108003.1 | NP_001101473.1 | 3 | 441 | ||
| XM_008764147.1 | XP_008762369.1 | 5 | 1008 | |||