siRNA Details
| Gene Symbol: Dnajb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108004.1 | NP_001101474.1 | 2, 3 | 265 | ||
| XM_006238070.2 | XP_006238132.1 | 2, 3 | 298 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dnajb5| Gene Symbol: Dnajb5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108004.1 | NP_001101474.1 | 2, 3 | 265 | ||
| XM_006238070.2 | XP_006238132.1 | 2, 3 | 298 | |||