siRNA Details
| Gene Symbol: Mast2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108005.1 | NP_001101475.1 | 24 | 3095 | ||
| XM_006238671.2 | XP_006238733.1 | 25 | 3281 | |||
| XM_006238672.2 | XP_006238734.1 | 24 | 3114 | |||
| XM_006238674.2 | XP_006238736.1 | 25 | 3107 | |||
| XM_006238675.2 | XP_006238737.1 | 24 | 3086 | |||

Mast2
Genomic Map