siRNA Details
| Gene Symbol: Vps13d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108006.1 | NP_001101476.1 | 19 | 4513 | ||
| XM_006239349.2 | XP_006239411.1 | 19 | 4508 | |||
| XM_006239350.2 | XP_006239412.1 | 19 | 4478 | |||
| XM_006239351.2 | XP_006239413.1 | 19 | 4475 | |||
| XM_006239352.2 | XP_006239414.1 | 19 | 4477 | |||
| XM_006239353.2 | XP_006239415.1 | 19 | 4477 | |||

Vps13d
Genomic Map