siRNA Details
| Gene Symbol: RGD1562284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134557.2 | NP_001128029.1 | 2 | 241 | ||
| XM_006239632.2 | XP_006239694.1 | 3 | 606 | |||
| GenBank mRNA | FQ228724.1 | - | 3 | 397 | ||
| FQ229106.1 | - | 3 | 400 | |||

RGD1562284
Genomic Map