siRNA Details
| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108008.1 | NP_001101478.1 | 10 | 969 | ||
| XM_006239681.2 | XP_006239743.1 | 11 | 1856 | |||
| XM_006239682.2 | XP_006239744.1 | 10 | 1823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eml4| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108008.1 | NP_001101478.1 | 10 | 969 | ||
| XM_006239681.2 | XP_006239743.1 | 11 | 1856 | |||
| XM_006239682.2 | XP_006239744.1 | 10 | 1823 | |||