siRNA Details
| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239681.2 | XP_006239743.1 | 7, 8 | 1516 | ||
| XM_006239682.2 | XP_006239744.1 | 6, 7 | 1483 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eml4| Gene Symbol: Eml4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239681.2 | XP_006239743.1 | 7, 8 | 1516 | ||
| XM_006239682.2 | XP_006239744.1 | 6, 7 | 1483 | |||