siRNA Details
| Gene Symbol: Lrpprc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008519.1 | NP_001008519.1 | 14, 15 | 1653 | ||
| GenBank mRNA | AY293808.1 | - | 14, 15 | 1653 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lrpprc| Gene Symbol: Lrpprc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008519.1 | NP_001008519.1 | 14, 15 | 1653 | ||
| GenBank mRNA | AY293808.1 | - | 14, 15 | 1653 | ||