siRNA Details
| Gene Symbol: Supt7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108010.2 | NP_001101480.1 | 4 | 1024 | ||
| GenBank mRNA | BC168944.1 | - | 4 | 1024 | 8371512 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Supt7l| Gene Symbol: Supt7l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108010.2 | NP_001101480.1 | 4 | 1024 | ||
| GenBank mRNA | BC168944.1 | - | 4 | 1024 | 8371512 | |