siRNA Details
| Gene Symbol: Smc6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108014.1 | NP_001101484.1 | 18 | 2234 | ||
| XM_006239898.2 | XP_006239960.1 | 18 | 2393 | |||
| GenBank mRNA | FQ230575.1 | - | 7 | 981 | ||
| FQ234676.1 | - | 7 | 980 | |||

Smc6
Genomic Map