siRNA Details
| Gene Symbol: Lpin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012111.1 | NP_001012111.1 | 16 | 2415 | ||
| XM_008764593.1 | XP_008762815.1 | 17 | 2382 | |||
| GenBank mRNA | BC083651.1 | - | 16 | 2415 | 7135110 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lpin1| Gene Symbol: Lpin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012111.1 | NP_001012111.1 | 16 | 2415 | ||
| XM_008764593.1 | XP_008762815.1 | 17 | 2382 | |||
| GenBank mRNA | BC083651.1 | - | 16 | 2415 | 7135110 | |