siRNA Details
| Gene Symbol: E2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100717.1 | NP_001094187.1 | 6 | 728 | ||
| XM_006239913.2 | XP_006239975.1 | 7 | 775 | |||
| GenBank mRNA | EV774540.1 | - | 3 | 297 | ||
| FQ216870.1 | - | 6 | 973 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E2f6| Gene Symbol: E2f6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100717.1 | NP_001094187.1 | 6 | 728 | ||
| XM_006239913.2 | XP_006239975.1 | 7 | 775 | |||
| GenBank mRNA | EV774540.1 | - | 3 | 297 | ||
| FQ216870.1 | - | 6 | 973 | |||