siRNA Details
| Gene Symbol: Cfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199378.3 | NP_955410.1 | 5 | 695 | ||
| GenBank mRNA | AB125856.1 | - | 5 | 575 | ||
| BC067246.1 | - | 5 | 699 | 6889499 | ||
| FQ212332.1 | - | 729 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cfdp1| Gene Symbol: Cfdp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199378.3 | NP_955410.1 | 5 | 695 | ||
| GenBank mRNA | AB125856.1 | - | 5 | 575 | ||
| BC067246.1 | - | 5 | 699 | 6889499 | ||
| FQ212332.1 | - | 729 | ||||