siRNA Details
| Gene Symbol: Cmpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108017.1 | NP_001101487.1 | 1, 2 | 678 | ||
| XM_006239938.2 | XP_006240000.1 | 2, 3 | 1365 | |||
| XM_006239939.2 | XP_006240001.1 | 2, 3 | 1365 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cmpk2| Gene Symbol: Cmpk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108017.1 | NP_001101487.1 | 1, 2 | 678 | ||
| XM_006239938.2 | XP_006240000.1 | 2, 3 | 1365 | |||
| XM_006239939.2 | XP_006240001.1 | 2, 3 | 1365 | |||