siRNA Details
| Gene Symbol: Hdac9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001200045.1 | NP_001186974.1 | 25 | 3377 | ||
| XM_008764616.1 | XP_008762838.1 | 24 | 3423 | |||
| GenBank mRNA | AB558551.1 | - | 25 | 3377 | ||
| FQ225774.1 | - | 20 | 2397 | |||

Hdac9
Genomic Map