siRNA Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199495.2 | NP_955789.2 | 5, 6 | 737 | ||
| XM_008767385.1 | XP_008765607.1 | 5, 6 | 749 | |||
| GenBank mRNA | BC061985.1 | - | 5, 6 | 564 | 5599238 | |
| BC093375.1 | - | 5, 6 | 696 | 7389276 | ||
| FQ209525.1 | - | 5, 6 | 739 | |||
| FQ215915.1 | - | 4, 5 | 454 | |||
| Gene Symbol: Ndufa10l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_182671.2 | NP_872612.1 | 1 | 699 | ||
| GenBank mRNA | AY260163.1 | - | 1 | 699 | ||

Ndufa10
Genomic Map