siRNA Details
| Gene Symbol: Ndufa10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199495.2 | NP_955789.2 | 5 | 668 | ||
| XM_008767385.1 | XP_008765607.1 | 5 | 680 | |||
| GenBank mRNA | BC061985.1 | - | 5 | 495 | 5599238 | |
| BC093375.1 | - | 5 | 627 | 7389276 | ||
| FQ215915.1 | - | 4 | 385 | |||

Ndufa10
Genomic Map