siRNA Details
Gene Symbol: Ndufa10l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182671.2 | NP_872612.1 | 1 | 841 | ||
GenBank mRNA | AY260163.1 | - | 1 | 841 |
Gene Symbol: Ndufa10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199495.2 | NP_955789.2 | 8 | 879 | ||
XM_008767385.1 | XP_008765607.1 | 8 | 891 | |||
GenBank mRNA | BC061985.1 | - | 8 | 706 | 5599238 | |
BC093375.1 | - | 8 | 838 | 7389276 | ||
FQ209525.1 | - | 8 | 881 | |||
FQ215915.1 | - | 7 | 596 |
