siRNA Details
| Gene Symbol: L3hypdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108031.1 | NP_001101501.1 | 3 | 1027 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

L3hypdh| Gene Symbol: L3hypdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108031.1 | NP_001101501.1 | 3 | 1027 | ||