siRNA Details
| Gene Symbol: Zbtb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004444.1 | NP_001004444.1 | 1 | 164 | ||
| XM_006240241.2 | XP_006240303.1 | 2 | 378 | |||
| XM_006240243.2 | XP_006240305.1 | 2 | 385 | |||
| XM_008764769.1 | XP_008762991.1 | 2 | 378 | |||
| GenBank mRNA | AY693992.1 | - | 1 | 164 | ||

Zbtb1
Genomic Map