siRNA Details
| Gene Symbol: Plekhh1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108036.2 | NP_001101506.2 | 28 | 4129 | ||
| XM_006240262.2 | XP_006240324.1 | 28 | 3995 | |||
| XM_006240263.2 | XP_006240325.1 | 28 | 4098 | |||
| XM_008764775.1 | XP_008762997.1 | 28 | 3895 | |||
| XM_008764776.1 | XP_008762998.1 | 28 | 3992 | |||
| XM_008764777.1 | XP_008762999.1 | 28 | 3964 | |||

Plekhh1
Genomic Map