siRNA Details
| Gene Symbol: Rdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108037.1 | NP_001101507.1 | 5 | 643 | ||
| XM_008764779.1 | XP_008763001.1 | 4 | 773 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rdh12| Gene Symbol: Rdh12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108037.1 | NP_001101507.1 | 5 | 643 | ||
| XM_008764779.1 | XP_008763001.1 | 4 | 773 | |||