siRNA Details
| Gene Symbol: Zfyve26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108038.1 | NP_001101508.1 | 31 | 5898 | ||
| XM_008764780.1 | XP_008763002.1 | 31 | 5940 | |||
| XM_008764781.1 | XP_008763003.1 | 31 | 5934 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfyve26| Gene Symbol: Zfyve26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108038.1 | NP_001101508.1 | 31 | 5898 | ||
| XM_008764780.1 | XP_008763002.1 | 31 | 5940 | |||
| XM_008764781.1 | XP_008763003.1 | 31 | 5934 | |||