siRNA Details
| Gene Symbol: Elmsan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240336.2 | XP_006240398.1 | 4 | 8147 | ||
| XM_008764790.1 | XP_008763012.1 | 3 | 8240 | |||
| XM_008764791.1 | XP_008763013.1 | 3 | 2009 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Elmsan1| Gene Symbol: Elmsan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240336.2 | XP_006240398.1 | 4 | 8147 | ||
| XM_008764790.1 | XP_008763012.1 | 3 | 8240 | |||
| XM_008764791.1 | XP_008763013.1 | 3 | 2009 | |||