siRNA Details
| Gene Symbol: Zfp410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108042.2 | NP_001101512.1 | 3 | 290 | ||
| XM_006240338.1 | XP_006240400.1 | 3 | 311 | |||
| XM_006240339.1 | XP_006240401.1 | 3 | 1222 | |||
| GenBank mRNA | BC168180.1 | - | 3 | 290 | 9100560 | |

Zfp410
Genomic Map