siRNA Details
| Gene Symbol: Fos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022197.2 | NP_071533.1 | 4 | 1199 | ||
| GenBank mRNA | FQ225996.1 | - | 3 | 852 | ||
| FQ228884.1 | - | 3 | 853 | |||
| X06769.1 | - | 1181 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fos| Gene Symbol: Fos | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022197.2 | NP_071533.1 | 4 | 1199 | ||
| GenBank mRNA | FQ225996.1 | - | 3 | 852 | ||
| FQ228884.1 | - | 3 | 853 | |||
| X06769.1 | - | 1181 | ||||