siRNA Details
| Gene Symbol: Flvcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199109.2 | NP_954540.2 | 3 | 1230 | ||
| GenBank mRNA | AY260573.1 | - | 3 | 982 | ||
| BC101899.1 | - | 3 | 1230 | 7932187 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Flvcr2| Gene Symbol: Flvcr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199109.2 | NP_954540.2 | 3 | 1230 | ||
| GenBank mRNA | AY260573.1 | - | 3 | 982 | ||
| BC101899.1 | - | 3 | 1230 | 7932187 | ||