siRNA Details
| Gene Symbol: Lrrc74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037783.2 | NP_001032872.2 | 8, 9 | 1302 | ||
| GenBank mRNA | BC083569.1 | - | 8, 9 | 1251 | ||
| BC098927.1 | - | 1314 | ||||
| BC127444.1 | - | 8, 9 | 1293 | 7135116 | ||

Lrrc74
Genomic Map