siRNA Details
| Gene Symbol: Irf2bpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012470.1 | NP_001012488.1 | 1 | 2200 | ||
| GenBank mRNA | BC098759.1 | - | 1 | 901 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Irf2bpl| Gene Symbol: Irf2bpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012470.1 | NP_001012488.1 | 1 | 2200 | ||
| GenBank mRNA | BC098759.1 | - | 1 | 901 | ||