siRNA Details
| Gene Symbol: LOC100910720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_008771481.1 | XP_008769703.1 | 859 | |||
| Gene Symbol: Ddx41 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108046.2 | NP_001101516.1 | 10 | 1014 | ||
| GenBank mRNA | BC166825.1 | - | 10 | 1014 | 8370850 | |

LOC100910720
Genomic Map