siRNA Details
| Gene Symbol: Foxn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108047.1 | NP_001101517.1 | 4 | 800 | ||
| XM_006240429.2 | XP_006240491.1 | 2 | 598 | |||
| XM_006240431.2 | XP_006240493.1 | 3 | 1436 | |||
| XM_008764800.1 | XP_008763022.1 | 3 | 2173 | |||
| XM_008764801.1 | XP_008763023.1 | 4 | 868 | |||
| XM_008764802.1 | XP_008763024.1 | 3 | 2173 | |||
| XM_008764803.1 | XP_008763025.1 | 3 | 2173 | |||

Foxn3
Genomic Map