siRNA Details
| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108048.1 | NP_001101518.1 | 10 | 1373 | ||
| XM_006240437.2 | XP_006240499.1 | 11 | 1556 | |||
| XM_006240438.2 | XP_006240500.1 | 10 | 1484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rps6ka5| Gene Symbol: Rps6ka5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108048.1 | NP_001101518.1 | 10 | 1373 | ||
| XM_006240437.2 | XP_006240499.1 | 11 | 1556 | |||
| XM_006240438.2 | XP_006240500.1 | 10 | 1484 | |||