siRNA Details
| Gene Symbol: LOC102553138 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240453.2 | XP_006240515.1 | 1 | 1061 | ||
| Gene Symbol: Gpr68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001108049.1 | NP_001101519.1 | 1 | 892 | ||
More Information
Set Membership: |
|

LOC102553138
Genomic Map